Gene Transcripts

Gene Symbol
Sppl3
Entrez Gene ID
360822
Gene Name
signal peptide peptidase like 3
Gene Aliases
RGD1562826, Sppl3-ps1
Chromosome Location
Chr. 12: 47276485 - 47360552 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751200.3 XP_003751248.1 3 224

View Full Product Details