Gene Transcripts

Gene Symbol
RGS11
Entrez Gene ID
8786
Gene Name
regulator of G-protein signaling 11
Gene Aliases
RS11
Chromosome Location
Chr. 16: 268301 - 275943 on Build GRCh38
UniGene ID
Hs.65756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286485.1 NP_001273414.1 8, 9 663
NM_003834.2 NP_003825.1 9, 10 707
NM_183337.2 NP_899180.1 9, 10 696
XM_011522719.2 XP_011521021.1 10, 11 790
XM_011522720.2 XP_011521022.1 10, 11 730
GenBank mRNA AB016929.1 - 8, 9 631
AF035153.1 - 9, 10 667
AF035154.1 - 9, 10 678
AK293564.1 - 9, 10 690
AK308972.1 - 8, 9 917
BC141803.1 - 9, 10 671 40147243

View Full Product Details