Gene Transcripts

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
Chromosome Location
Chr. 13: 114234845 - 114272675 on Build GRCh38
UniGene ID
Hs.374127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078645.1 NP_001072113.1 14, 15 1513
NM_001318517.1 NP_001305446.1 14, 15 1510
NM_001318518.1 NP_001305447.1 13, 14 1357
NM_003903.3 NP_003894.3 14, 15 1513
XM_005266206.1 XP_005266263.1 14, 15 1555
XM_006719992.1 XP_006720055.1 14, 15 1500
XM_006719993.1 XP_006720056.1 13, 14 1448
XM_011534851.1 XP_011533153.1 13, 14 1402
XM_011534853.1 XP_011533155.1 14, 15 1555
XM_017020830.1 XP_016876319.1 13, 14 1402
XM_017020831.1 XP_016876320.1 14, 15 1555
XM_017020832.1 XP_016876321.1 13, 14 1448
XM_017020833.1 XP_016876322.1 12, 13 1133
GenBank mRNA AF164598.1 - 13, 14 1163
BC010875.2 - 14, 15 1457 3879150
BC017244.1 - 14, 15 1423 4877392
U18291.1 - 15, 16 1336

View Full Product Details