Gene Transcripts

Gene Symbol
Dmrtc1c1
Entrez Gene ID
363483
Gene Name
DMRT-like family C1c1
Gene Aliases
Dmrtc1c, Dmrtc1c2
Chromosome Location
Chr. X: 72684315 - 72691535 on Build Rnor_6.0
UniGene ID
Rn.30139
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014222.2 NP_001014244.2 4, 5 620
GenBank mRNA BC079417.1 - 4, 5 606
CV112810.1 - 4, 5 620

View Full Product Details