Gene Transcripts

Gene Symbol
Iars2
Entrez Gene ID
364070
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
RGD1311857
Chromosome Location
Chr. 13: 103231214 - 103265018 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769904.1 XP_008768126.1 5 825

View Full Product Details