Gene Transcripts

Gene Symbol
Vom2r70
Entrez Gene ID
364086
Gene Name
vomeronasal 2 receptor, 70
Gene Aliases
RGD1562647
Chromosome Location
Chr. 14: 308569 - 338157 on Build Rnor_6.0
UniGene ID
Rn.217631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099477.1 NP_001092947.1 6 2095

View Full Product Details