Gene Transcripts

Gene Symbol
Lsm1
Entrez Gene ID
364624
Gene Name
LSM1, U6 small nuclear RNA associated
Gene Aliases
Chromosome Location
Chr. 16: 71046475 - 71057883 on Build Rnor_6.0
UniGene ID
Rn.128606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108876.1 NP_001102346.1 3 507
GenBank mRNA FQ216033.1 - 3 277
FQ220497.1 - 3 375

View Full Product Details