Gene Transcripts

Gene Symbol
Bves
Entrez Gene ID
365603
Gene Name
blood vessel epicardial substance
Gene Aliases
Popdc1
Chromosome Location
Chr. 20: 50439885 - 50474678 on Build Rnor_6.0
UniGene ID
Rn.41396
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077590.1 NP_001071058.1 3 540
GenBank mRNA AY786529.2 - 3 540

View Full Product Details