Gene Transcripts

Gene Symbol
Arsk
Entrez Gene ID
365619
Gene Name
arylsulfatase family, member K
Gene Aliases
RGD1310182
Chromosome Location
Chr. 2: 2835568 - 2891059 on Build Rnor_6.0
UniGene ID
Rn.202360
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047917.1 NP_001041382.1 5 796

View Full Product Details