Gene Transcripts
- Gene Symbol
- ATP1A2
- Entrez Gene ID
- 477
- Gene Name
- ATPase Na+/K+ transporting subunit alpha 2
- Gene Aliases
- FHM2, MHP2
- Chromosome Location
- Chr. 1: 160115730 - 160143591 on Build GRCh38
- UniGene ID
- Hs.34114
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000702.3 |
NP_000693.1
|
4, 5 |
508 |
|
GenBank mRNA |
AB018321.2 |
- |
4, 5 |
465 |
|
AK091617.1 |
- |
4, 5 |
479 |
|
AK314296.1 |
- |
4, 5 |
479 |
|
AL831991.1 |
- |
3, 4 |
297 |
|
AL831997.1 |
- |
3, 4 |
323 |
|
AY946014.1 |
- |
1, 2 |
131 |
|
BC052271.1 |
- |
4, 5 |
459 |
6672133 |
DA221602.1 |
- |
4, 5 |
508 |
|
KU177927.1 |
- |
3, 4 |
376 |
|
KU177928.1 |
- |
3, 4 |
376 |
|
View Full Product Details