Gene Transcripts

Gene Symbol
USP6
Entrez Gene ID
9098
Gene Name
ubiquitin specific peptidase 6
Gene Aliases
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short
Chromosome Location
Chr. 17: 5116438 - 5175032 on Build GRCh38
UniGene ID
Hs.448851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304284.1 NP_001291213.1 24 3875
NM_004505.3 NP_004496.2 16 3344
XM_011524050.1 XP_011522352.1 20 4093
XM_011524051.2 XP_011522353.1 22 3188
XM_011524052.2 XP_011522354.1 21 3302
XM_011524053.2 XP_011522355.1 22 3215
XM_011524054.2 XP_011522356.1 22 3182
XM_011524055.2 XP_011522357.1 20 3109
XM_011524056.2 XP_011522358.1 17 3062
XM_011524057.2 XP_011522359.1 20 3225
XM_011524058.2 XP_011522360.1 21 3291
XM_011524059.2 XP_011522361.1 21 3291
XM_017025290.1 XP_016880779.1 19 3931
GenBank mRNA AB449915.1 - 15 1645
AY143550.1 - 16 3344
BX647719.1 - 24 3876
X63546.1 - 16 3341
X63547.1 - 17 3552

View Full Product Details