Gene Transcripts
- Gene Symbol
- DGKI
- Entrez Gene ID
- 9162
- Gene Name
- diacylglycerol kinase iota
- Gene Aliases
- DGK-IOTA
- Chromosome Location
- Chr. 7: 137384283 - 137846864 on Build GRCh38
- UniGene ID
- Hs.242947
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321708.1 |
NP_001308637.1
|
30, 31 |
2933 |
|
NM_001321709.1 |
NP_001308638.1
|
31, 32 |
2711 |
|
NM_001321710.1 |
NP_001308639.1
|
30, 31 |
2830 |
|
NM_004717.3 |
NP_004708.1
|
31, 32 |
2957 |
|
XM_011516684.2 |
XP_011514986.2
|
24, 25 |
10177 |
|
XM_017012780.1 |
XP_016868269.1
|
31, 32 |
2994 |
|
XM_017012781.1 |
XP_016868270.1
|
31, 32 |
2978 |
|
XM_017012782.1 |
XP_016868271.1
|
30, 31 |
2879 |
|
XM_017012783.1 |
XP_016868272.1
|
30, 31 |
2870 |
|
XM_017012784.1 |
XP_016868273.1
|
29, 30 |
2804 |
|
XM_017012785.1 |
XP_016868274.1
|
29, 30 |
3181 |
|
XM_017012786.1 |
XP_016868275.1
|
31, 32 |
2681 |
|
XM_017012787.1 |
XP_016868276.1
|
30, 31 |
2546 |
|
XM_017012788.1 |
XP_016868277.1
|
31, 32 |
2664 |
|
XM_017012789.1 |
XP_016868278.1
|
17, 18 |
1390 |
|
GenBank mRNA |
AB209167.1 |
- |
30, 31 |
2502 |
|
AK091081.1 |
- |
4, 5 |
577 |
|
View Full Product Details