Gene Transcripts

Gene Symbol
Btnl7
Entrez Gene ID
406159
Gene Name
butyrophilin-like 7
Gene Aliases
Chromosome Location
Chr. 20: 2757008 - 2767240 on Build Rnor_6.0
UniGene ID
Rn.17843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212488.1 NP_997653.1 884
XM_008772737.1 XP_008770959.1 884

Gene Symbol
LOC100911938
Entrez Gene ID
100911938
Gene Name
butyrophilin subfamily 3 member A3-like
Gene Aliases
Chromosome Location
Chr. 20: 2790805 - 2823562 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772767.1 XP_008770989.1 884

Gene Symbol
LOC100911983
Entrez Gene ID
100911983
Gene Name
butyrophilin subfamily 3 member A3-like
Gene Aliases
Chromosome Location
Chr. 20: 2909474 - 2925869 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772783.1 XP_008771005.1 5 1073

View Full Product Details