Gene Transcripts

Gene Symbol
Ly49si3
Entrez Gene ID
494209
Gene Name
immunoreceptor Ly49si3
Gene Aliases
Chromosome Location
Chr. 4: 163986547 - 164028470 on Build Rnor_6.0
UniGene ID
Rn.134171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009919.1 NP_001009919.1 3 390
GenBank mRNA AY653730.1 - 3 390

View Full Product Details