Gene Transcripts

Gene Symbol
Vom1r10
Entrez Gene ID
494302
Gene Name
vomeronasal 1 receptor 10
Gene Aliases
V1re21
Chromosome Location
Chr. 1: 60280451 - 60281386 on Build Rnor_6.0
UniGene ID
Rn.129456
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008958.1 NP_001008958.1 1 537
GenBank mRNA AY510358.1 - 1 537

View Full Product Details