Gene Transcripts

Gene Symbol
Vom1r28
Entrez Gene ID
494308
Gene Name
vomeronasal 1 receptor 28
Gene Aliases
V1re16
Chromosome Location
Chr. 1: 68176904 - 68177836 on Build Rnor_6.0
UniGene ID
Rn.129438
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008966.1 NP_001008966.1 1 199
GenBank mRNA AY510353.1 - 1 199

Gene Symbol
LOC100911184
Entrez Gene ID
100911184
Gene Name
vomeronasal type-1 receptor 4-like
Gene Aliases
Chromosome Location
Chr. 1: 68239224 - 68240407 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228164.2 XP_006228226.1 1 289

Gene Symbol
LOC103691019
Entrez Gene ID
103691019
Gene Name
vomeronasal type-1 receptor 4-like
Gene Aliases
Chromosome Location
Chr. 1: 63201271 - 63202203 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758872.1 XP_008757094.1 1 199

Gene Symbol
Vom1r109
Entrez Gene ID
494228
Gene Name
vomeronasal 1 receptor 109
Gene Aliases
V1re17, Vom1r-ps39
Chromosome Location
Chr. 1: 63263722 - 63264582 on Build Rnor_6.0
UniGene ID
Rn.144524
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009509.1 NP_001009509.1 1 199
GenBank mRNA AY510360.1 - 1 199

View Full Product Details