Gene Transcripts

Gene Symbol
Vom1r93
Entrez Gene ID
497787
Gene Name
vomeronasal 1 receptor 93
Gene Aliases
V1rb5
Chromosome Location
Chr. 4: 121771681 - 121779354 on Build Rnor_6.0
UniGene ID
Rn.10142
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008971.1 NP_001008971.1 1 453
XM_006236848.1 XP_006236910.1 3 789
GenBank mRNA AY510293.1 - 1 453
U36896.1 - 3 803

View Full Product Details