Gene Transcripts

Gene Symbol
PTTG1
Entrez Gene ID
9232
Gene Name
pituitary tumor-transforming 1
Gene Aliases
EAP1, HPTTG, PTTG, TUTR1
Chromosome Location
Chr. 5: 160421807 - 160428744 on Build GRCh38
UniGene ID
Hs.350966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282382.1 NP_001269311.1 1 458
NM_001282383.1 NP_001269312.1 2 312
NM_004219.3 NP_004210.1 2 151
GenBank mRNA AF062649.1 - 2 125
AF075242.1 - 1 75
AF095287.1 - 2 91
AJ223953.1 - 1 160
AK312209.1 - 2 129
BC101834.1 - 2 84 8069340
BC108894.1 - 1 55
BC108895.1 - 1 55
BC112071.1 - 2 84 8327539
BC113842.1 - 1 55
BC113896.1 - 1 55
BC128193.1 - 1 55 40113885
BC128194.1 - 1 55 40113886
BI827526.1 - 1 458
BU182149.1 - 2 307
CR457135.1 - 1 55
CR541685.1 - 1 55
HY006026.1 - 2 152

View Full Product Details