Gene Transcripts

Gene Symbol
Lmod3
Entrez Gene ID
500267
Gene Name
leiomodin 3 (fetal)
Gene Aliases
RGD1564924
Chromosome Location
Chr. 4: 129604378 - 129619140 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236972.2 XP_006237034.1 3 598
XM_006236973.2 XP_006237035.1 3 485
XM_575617.7 XP_575617.4 2 590

View Full Product Details