Gene Transcripts

Gene Symbol
CDY2A
Entrez Gene ID
9426
Gene Name
chromodomain protein, Y-linked, 2A
Gene Aliases
CDY, CDY2
Chromosome Location
Chr. Y: 18025787 - 18027746 on Build GRCh38
UniGene ID
Hs.251375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004825.2 NP_004816.1 1 1190
GenBank mRNA AF080598.1 - 1 1190
AK292233.1 - 1 1163

Gene Symbol
CDY1
Entrez Gene ID
9085
Gene Name
chromodomain protein, Y-linked, 1
Gene Aliases
CDY, CDY1A
Chromosome Location
Chr. Y: 25622115 - 25625511 on Build GRCh38
UniGene ID
Hs.159281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004680.2 NP_004671.1 1 1185
NM_170723.1 NP_733841.1 1 1185
XM_011531512.1 XP_011529814.1 1 1187
GenBank mRNA AF000981.1 - 1 1140
AF080597.1 - 1 1140
AK292375.1 - 1 1207
BC033041.1 - 1 1190
BC132955.1 - 1 954 40146977

Gene Symbol
CDY1B
Entrez Gene ID
253175
Gene Name
chromodomain protein, Y-linked, 1B
Gene Aliases
CDY
Chromosome Location
Chr. Y: 24044620 - 24048016 on Build GRCh38
UniGene ID
Hs.562095
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003894.1 NP_001003894.1 1 1185
NM_001003895.1 NP_001003895.1 1 1185
XM_011531484.1 XP_011529786.1 1 1187
GenBank mRNA AK292318.1 - 1 1158
BC132929.1 - 1 954 40146951

Gene Symbol
CDY2B
Entrez Gene ID
203611
Gene Name
chromodomain protein, Y-linked, 2B
Gene Aliases
CDY
Chromosome Location
Chr. Y: 17878260 - 17880219 on Build GRCh38
UniGene ID
Hs.532657
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001722.1 NP_001001722.1 1 1189
GenBank mRNA BC069087.1 - 1 1189
BC130426.1 - 1 958 40146457
BC130428.1 - 1 958 40146459

View Full Product Details