Gene Transcripts

Gene Symbol
Vom2r68
Entrez Gene ID
501881
Gene Name
vomeronasal 2 receptor, 68
Gene Aliases
RGD1560816
Chromosome Location
Chr. 14: 1106463 - 1113322 on Build Rnor_6.0
UniGene ID
Rn.217937
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099481.1 NP_001092951.1 6 1798

View Full Product Details