Gene Transcripts

Gene Symbol
Klri2
Entrez Gene ID
503650
Gene Name
killer cell lectin-like receptor family I member 2
Gene Aliases
Chromosome Location
Chr. 4: 163561525 - 163571116 on Build Rnor_6.0
UniGene ID
Rn.160081
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012648.1 NP_001012666.1 6 702
XM_008763327.1 XP_008761549.1 5 946
GenBank mRNA AY324871.1 - 6 702

View Full Product Details