Gene Transcripts

Gene Symbol
Ube2d2
Entrez Gene ID
641452
Gene Name
ubiquitin-conjugating enzyme E2D 2
Gene Aliases
E217kB, Ubc2e
Chromosome Location
Chr. 18: 28561957 - 28603335 on Build Rnor_6.0
UniGene ID
Rn.7390
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037292.1 NP_001032369.1 157
GenBank mRNA U13176.1 - 157

View Full Product Details