Gene Transcripts

Gene Symbol
Dnd1
Entrez Gene ID
679841
Gene Name
DND microRNA-mediated repression inhibitor 1
Gene Aliases
Chromosome Location
Chr. 18: 29608588 - 29611212 on Build Rnor_6.0
UniGene ID
Rn.160920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109379.1 NP_001102849.1 4 746

View Full Product Details