Gene Transcripts

Gene Symbol
Tcf24
Entrez Gene ID
680678
Gene Name
transcription factor 24
Gene Aliases
Chromosome Location
Chr. 5: 8957110 - 8961523 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763517.1 XP_008761739.1 4 587
XM_008763519.1 XP_008761741.1 3 553

View Full Product Details