Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004905.2 | NP_004896.1 | 5 | 667 | |
GenBank mRNA | AK095940.1 | - | 1 | 881 | |
AK289352.1 | - | 5 | 665 | ||
AK300680.1 | - | 6 | 564 | ||
BC035857.1 | - | 5 | 667 | 5767682 | |
BC053550.1 | - | 5 | 617 | 6013037 | |
D14662.1 | - | 5 | 659 | ||
EU794652.1 | - | 5 | 667 |