Gene Transcripts

Gene Symbol
Vom2r15
Entrez Gene ID
690680
Gene Name
vomeronasal 2 receptor, 15
Gene Aliases
Chromosome Location
Chr. 1: 62665651 - 62681471 on Build Rnor_6.0
UniGene ID
Rn.217648
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099490.1 NP_001092960.1 1852

View Full Product Details