Gene Transcripts

Gene Symbol
SH3PXD2A
Entrez Gene ID
9644
Gene Name
SH3 and PX domains 2A
Gene Aliases
FISH, SH3MD1, TKS5
Chromosome Location
Chr. 10: 103594027 - 103855464 on Build GRCh38
UniGene ID
Hs.678727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014631.2 NP_055446.2 10 965
XM_005270294.4 XP_005270351.1 11 1098
XM_005270295.4 XP_005270352.1 10 1052
XM_005270297.3 XP_005270354.1 5 2329
XM_005270298.4 XP_005270355.1 5 1132
XM_006718079.3 XP_006718142.1 12 1195
XM_006718080.3 XP_006718143.1 6 1226
XM_011540385.2 XP_011538687.1 11 1149
XM_011540386.2 XP_011538688.1 6 2425
XM_011540387.2 XP_011538689.1 2 4071
XM_017016969.1 XP_016872458.1 11 1116
XM_017016970.1 XP_016872459.1 10 932
GenBank mRNA AB007878.1 - 8 715
AK056469.1 - 9 688
AK294454.1 - 5 505
AK294736.1 - 5 543
AK295314.1 - 5 582
AK295660.1 - 5 609
AK302015.1 - 8 721
AK304828.1 - 5 521
AK315983.1 - 5 548

View Full Product Details