Gene Transcripts

Gene Symbol
Ppapdc1b
Entrez Gene ID
680466
Gene Name
phosphatidic acid phosphatase type 2 domain containing 1B
Gene Aliases
Chromosome Location
Chr. 16: 71121175 - 71125794 on Build Rnor_6.0
UniGene ID
Rn.199008
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109411.1 NP_001102881.2 4 317
XM_008771367.1 XP_008769589.1 4 795

View Full Product Details