Gene Transcripts

Gene Symbol
Chchd7
Entrez Gene ID
684258
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 7
Gene Aliases
Chromosome Location
Chr. 5: 16843246 - 16847809 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749892.3 XP_003749940.1 4 506
XM_003749893.2 XP_003749941.1 4 345
XM_008763563.1 XP_008761785.1 4 568

View Full Product Details