Gene Transcripts

Gene Symbol
Sys1
Entrez Gene ID
685079
Gene Name
Sys1 golgi trafficking protein
Gene Aliases
Chromosome Location
Chr. 3: 160908770 - 160921327 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001062200.5 XP_001062200.2 3 515
XM_008762556.1 XP_008760778.1 2 419
XM_008762557.1 XP_008760779.1 2 428

View Full Product Details