Gene Transcripts

Gene Symbol
Lrrcc1
Entrez Gene ID
266808
Gene Name
leucine rich repeat and coiled-coil centrosomal protein 1
Gene Aliases
Sap2
Chromosome Location
Chr. 2: 88384796 - 88414012 on Build Rnor_6.0
UniGene ID
Rn.212242
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100645.2 NP_001094115.2 4 442
GenBank mRNA CB547116.1 - 4 388

View Full Product Details