Gene Transcripts

Gene Symbol
Or7e24
Entrez Gene ID
690849
Gene Name
olfactory receptor, family 7, subfamily E, member 24
Gene Aliases
Chromosome Location
Chr. 8: 21214143 - 21224737 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766006.1 XP_008764228.1 2 548

View Full Product Details