Gene Transcripts

Gene Symbol
Skint1
Entrez Gene ID
689801
Gene Name
selection and upkeep of intraepithelial T cells 1
Gene Aliases
Chromosome Location
Chr. 5: 131915491 - 131943133 on Build Rnor_6.0
UniGene ID
Rn.220367
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135916.1 NP_001129388.1 3 443
GenBank mRNA EU650232.1 - 3 443

View Full Product Details