Gene Transcripts

Gene Symbol
Prima1
Entrez Gene ID
690195
Gene Name
proline rich membrane anchor 1
Gene Aliases
PRiMA, Prima1l
Chromosome Location
Chr. 6: 127075954 - 127128014 on Build Rnor_6.0
UniGene ID
Rn.152596
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108721.2 NP_001102191.2 5 863
XM_006240495.2 XP_006240557.1 5 1306
XM_008764849.1 XP_008763071.1 5 936

View Full Product Details