Gene Transcripts

Gene Symbol
Vom2r37
Entrez Gene ID
690356
Gene Name
vomeronasal 2 receptor, 37
Gene Aliases
Chromosome Location
Chr. 1: 98989754 - 99135977 on Build Rnor_6.0
UniGene ID
Rn.217935
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099487.1 NP_001092957.1 2 975

View Full Product Details