Gene Transcripts

Gene Symbol
Kpna7
Entrez Gene ID
690504
Gene Name
karyopherin alpha 7
Gene Aliases
Chromosome Location
Chr. 12: 11347643 - 11384349 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001074637.3 XP_001074637.3 3, 4 286

View Full Product Details