Gene Transcripts

Gene Symbol
Vom2r33
Entrez Gene ID
691763
Gene Name
vomeronasal 2 receptor, 33
Gene Aliases
Chromosome Location
Chr. 1: 65937820 - 65952790 on Build Rnor_6.0
UniGene ID
Rn.217933
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099497.1 NP_001092967.1 1, 2 281

View Full Product Details