Gene Transcripts

Gene Symbol
DHDH
Entrez Gene ID
27294
Gene Name
dihydrodiol dehydrogenase
Gene Aliases
2DD, HUM2DD
Chromosome Location
Chr. 19: 48932790 - 48944969 on Build GRCh38
UniGene ID
Hs.631555
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014475.3 NP_055290.1 3 320
XM_017026598.1 XP_016882087.1 3 880
GenBank mRNA AB021933.1 - 3 280
BC032730.1 - 3 331 5530225

View Full Product Details