Gene Transcripts
- Gene Symbol
- TBC1D26
- Entrez Gene ID
- 353149
- Gene Name
- TBC1 domain family member 26
- Gene Aliases
- Chromosome Location
- Chr. 17: 15732277 - 15744784 on Build GRCh38
- UniGene ID
- Hs.434123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK292544.1 |
- |
9 |
875 |
|
AK315847.1 |
- |
9 |
875 |
|
- Gene Symbol
- TBC1D28
- Entrez Gene ID
- 254272
- Gene Name
- TBC1 domain family member 28
- Gene Aliases
- Chromosome Location
- Chr. 17: 18634434 - 18644431 on Build GRCh38
- UniGene ID
- Hs.434123
- Species
- Homo sapiens
View Full Product Details