Gene Transcripts

Gene Symbol
RELL2
Entrez Gene ID
285613
Gene Name
RELT like 2
Gene Aliases
C5orf16
Chromosome Location
Chr. 5: 141636946 - 141641077 on Build GRCh38
UniGene ID
Hs.120094
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130029.1 NP_001123501.1 7, 8 1757
NM_173828.4 NP_776189.3 6, 7 2185
XM_011537625.2 XP_011535927.1 5, 6 2305
GenBank mRNA AK054889.1 - 4, 5 1932
AK075064.1 - 6, 7 1012
AY358163.1 - 6, 7 2121
BC063469.1 - 7, 8 1226 5211730

View Full Product Details