Gene Transcripts

Gene Symbol
C2orf27A
Entrez Gene ID
29798
Gene Name
chromosome 2 open reading frame 27A
Gene Aliases
C2orf27, C2orf27B
Chromosome Location
Chr. 2: 131722491 - 131767404 on Build GRCh38
UniGene ID
Hs.635289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013310.3 NP_037442.3 4 979
GenBank mRNA AF038169.1 - 4 979
BC071972.1 - 4 647
BC093716.1 - 2 599 7939561
BC112189.1 - 2 599 8327657

Gene Symbol
C2orf27B
Entrez Gene ID
408029
Gene Name
chromosome 2 open reading frame 27B
Gene Aliases
Chromosome Location
Chr. 2: 131794961 - 131801661 on Build GRCh38
UniGene ID
Hs.145567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_214461.2 NP_999626.1 3 1085
GenBank mRNA BC043584.1 - 3 1085

Gene Symbol
LOC105372731
Entrez Gene ID
105372731
Gene Name
CMT1A duplicated region transcript 15 protein-like protein
Gene Aliases
Chromosome Location
Chr. 21: 9026613 - 9036543 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017028519.1 XP_016884008.1 2 796

View Full Product Details