Gene Transcripts

Gene Symbol
RPS29
Entrez Gene ID
6235
Gene Name
ribosomal protein S29
Gene Aliases
DBA13, S29
Chromosome Location
Chr. 14: 49576672 - 49586416 on Build GRCh38
UniGene ID
Hs.156367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030001.2 NP_001025172.1 2 162
NM_001032.4 NP_001023.1 2 162
GenBank mRNA AK026649.1 - 1 122
AK307833.1 - 2 123
BC015974.1 - 2 126 4105424
BC032813.1 - 1 97 5217083
BC035313.1 - 97 3868721
BF128896.1 - 2 69
BQ433530.1 - 2 106
BU942244.1 - 2 165
L31610.1 - 2 103
U14973.1 - 2 122

View Full Product Details