Gene Transcripts
- Gene Symbol
- FAM72A
- Entrez Gene ID
- 729533
- Gene Name
- family with sequence similarity 72 member A
- Gene Aliases
- LMPIP, Ugene
- Chromosome Location
- Chr. 1: 206186179 - 206204918 on Build GRCh38
- UniGene ID
- Hs.339665
- Species
- Homo sapiens
- Gene Symbol
- FAM72B
- Entrez Gene ID
- 653820
- Gene Name
- family with sequence similarity 72 member B
- Gene Aliases
- p17
- Chromosome Location
- Chr. 1: 121167646 - 121186896 on Build GRCh38
- UniGene ID
- Hs.339665
- Species
- Homo sapiens
- Gene Symbol
- FAM72C
- Entrez Gene ID
- 554282
- Gene Name
- family with sequence similarity 72 member C
- Gene Aliases
- Chromosome Location
- Chr. 1: 143955364 - 143974219 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- FAM72D
- Entrez Gene ID
- 728833
- Gene Name
- family with sequence similarity 72 member D
- Gene Aliases
- GCUD2
- Chromosome Location
- Chr. 1: 145093805 - 145112696 on Build GRCh38
- UniGene ID
- Hs.535577
- Species
- Homo sapiens
View Full Product Details