Gene Transcripts
- Gene Symbol
- NUTM2A
- Entrez Gene ID
- 728118
- Gene Name
- NUT family member 2A
- Gene Aliases
- FAM22A
- Chromosome Location
- Chr. 10: 87221899 - 87235372 on Build GRCh38
- UniGene ID
- Hs.710565
- Species
- Homo sapiens
- Gene Symbol
- NUTM2B
- Entrez Gene ID
- 729262
- Gene Name
- NUT family member 2B
- Gene Aliases
- FAM22B, bA119F19.1
- Chromosome Location
- Chr. 10: 79703227 - 79712757 on Build GRCh38
- UniGene ID
- Hs.742755
- Species
- Homo sapiens
- Gene Symbol
- NUTM2D
- Entrez Gene ID
- 728130
- Gene Name
- NUT family member 2D
- Gene Aliases
- FAM22D
- Chromosome Location
- Chr. 10: 87357720 - 87370695 on Build GRCh38
- UniGene ID
- Hs.701044
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB095940.1 |
- |
2 |
997 |
|
View Full Product Details