Gene Transcripts

Gene Symbol
DHRS2
Entrez Gene ID
10202
Gene Name
dehydrogenase/reductase 2
Gene Aliases
HEP27, SDR25C1
Chromosome Location
Chr. 14: 23630115 - 23645639 on Build GRCh38
UniGene ID
Hs.272499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005794.3 NP_005785.1 4 858
NM_182908.4 NP_878912.1 4 858
XM_005267249.1 XP_005267306.1 4 570
XM_006720001.3 XP_006720064.1 4 533
XM_011536338.1 XP_011534640.1 3 499
XM_011536339.2 XP_011534641.1 3 349
GenBank mRNA AK222857.1 - 4 858
BC002786.2 - 4 849 3614599
BC007339.2 - 3 384
U31875.1 - 4 844

View Full Product Details