Gene Transcripts

Gene Symbol
LRRC23
Entrez Gene ID
10233
Gene Name
leucine rich repeat containing 23
Gene Aliases
LRPB7
Chromosome Location
Chr. 12: 6904733 - 6914242 on Build GRCh38
UniGene ID
Hs.155586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135217.1 NP_001128689.1 4 832
NM_006992.3 NP_008923.1 4 660
NM_201650.2 NP_964013.1 4 660
GenBank mRNA AK223535.1 - 4 594
AK292291.1 - 4 595
AK292367.1 - 4 660
AY834277.1 - 3 440
BC029858.1 - 4 633 5170643
U72508.1 - 4 552
U72509.1 - 2 267
U72510.1 - 3 442

View Full Product Details