Gene Transcripts

Gene Symbol
MUC3A
Entrez Gene ID
4584
Gene Name
mucin 3A, cell surface associated
Gene Aliases
MUC-3A, MUC3
Chromosome Location
Chr. 7: 100949054 - 100968347 on Build GRCh38
UniGene ID
Hs.703577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005960.1 NP_005951.1 2 8684
XM_011516246.2 XP_011514548.1 3 9013
XM_011516247.2 XP_011514549.1 3 8963
XM_011516249.2 XP_011514551.1 3 8814
XM_011516250.2 XP_011514552.1 3 8302
XM_017012231.1 XP_016867720.1 3 8510
GenBank mRNA AB038784.1 - 1 1141
AF007194.1 - 3 2162

View Full Product Details