Gene Transcripts

Gene Symbol
Pgd
Entrez Gene ID
110208
Gene Name
phosphogluconate dehydrogenase
Gene Aliases
0610042A05Rik, AU019875, C78335
Chromosome Location
Chr. 4: 149149985 - 149166707 on Build GRCm38
UniGene ID
Mm.252080
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081274.1 NP_001074743.1 3 277
GenBank mRNA AK002894.1 - 3 239
AK077110.1 - 3 239
AK145602.1 - 3 241
AK150210.1 - 3 240
AK150490.1 - 3 236
AK151380.1 - 3 236
AK151993.1 - 3 236
AK152758.1 - 3 236
AK152894.1 - 3 236
AK153409.1 - 3 233
AK155027.1 - 3 236
AK161869.1 - 3 235
AK166733.1 - 3 231
AK166947.1 - 3 213
AK167215.1 - 3 231
AK168251.1 - 3 236
BC008646.1 - 3 208 3155147
BC011329.1 - 3 226 3984654
BC014793.1 - 3 245 3155100

View Full Product Details