Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007750.2 | NP_031776.1 | 2 | 343 | |
GenBank mRNA | AK002218.1 | - | 2 | 303 | |
AK002524.1 | - | 2 | 259 | ||
AK008594.1 | - | 2 | 294 | ||
BC120708.1 | - | 2 | 264 | 40129631 | |
BC120710.1 | - | 2 | 264 | 40129633 | |
U37721.1 | - | 2 | 264 |